Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mdh2
Entrez Gene ID
42185
Gene Name
Malate dehydrogenase 2
Gene Aliases
CG 7998, CG7998, Dmel\CG7998, Dmel_CG7998, MDH-2, MDH2, Mdh, l(3)psg7, mMdh, mdh2, psg7
Chromosome Location
Chr.3R: 18226396 - 18228718 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262559
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_142439.3 NP_650696.1 3 - 4 578 68
GenBank mRNA AY119152.1 - 3 - 4 575 68
BT029274.1 - - 569 68

View Full Product Details