Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Mekk1
- Entrez Gene ID
-
42253
- Gene Name
- CG7717-PB
- Gene Aliases
-
CG7717, D-MEKK1, D-Mekk1, DmMEKK1, Dmel\CG7717, Dmel_CG7717, MEKK, MEKK1, MEKK4, Mekk1/4, dMEKK, dMEKK1, dMEKK4, dMekk1, mekk1, mekk1a, mekk1b
- Chromosome Location
-
Chr.3R: 18733106 - 18743594 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0024329
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142493.3 |
NP_650750.4 |
1 - 2 |
801 |
95 |
View Full Product Details