Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- trem
- Entrez Gene ID
-
42392
- Gene Name
- trade embargo
- Gene Aliases
-
CG4413, Dmel\CG4413, Dmel_CG4413
- Chromosome Location
-
Chr.3R: 20029560 - 20031309 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038767
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142604.2 |
NP_650861.1 |
3 - 4 |
1136 |
59 |
View Full Product Details