Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gfrl
Entrez Gene ID
3346162
Gene Name
Glial cell line-derived neurotrophic family receptor-like
Gene Aliases
BcDNA:GH26447, CG17203, CG17204, CG17205, CG17208, CG33518, CG34118, CG34119, CG42401, CG42625, CG42801, CG43224, CG4342, DmGfrl, Dmel\CG43224, Dmel_CG43224, dGFRLA, dGFRLAb, mun
Chromosome Location
Chr.3R: 20378458 - 20484588 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262869
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014642.5 NP_001014642.4 13 - 14 3573 70
NM_001260277.2 NP_001247206.1 13 - 14 2936 70
NM_001260278.2 NP_001247207.1 11 - 12 2862 70
NM_001260279.2 NP_001247208.1 14 - 15 3906 70
NM_001260280.2 NP_001247209.1 12 - 13 2617 70
NM_001260281.2 NP_001247210.1 13 - 14 2968 70
NM_001275827.2 NP_001262756.1 14 - 15 3906 70
NM_001300494.1 NP_001287423.1 11 - 12 2239 70
NM_001300495.1 NP_001287424.1 14 - 15 3906 70

View Full Product Details