Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DPCoAC
Entrez Gene ID
42429
Gene Name
Dephosphocoenzyme A carrier
Gene Aliases
CG4241, Dmel\CG4241, Dmel_CG4241, att, att-ORFA, dPCoAC
Chromosome Location
Chr.3R: 20552385 - 20557667 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0067783
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300499.1 NP_001287428.1 2 - 3 475 57
NM_001300501.1 NP_001287430.1 3 - 4 666 57
NM_142634.3 NP_650891.1 2 - 3 475 57
NM_206522.2 NP_996244.1 2 - 3 475 57
GenBank mRNA BT021264.1 - 2 - 3 462 57

View Full Product Details