Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrrk
Entrez Gene ID
42447
Gene Name
Leucine-rich repeat kinase
Gene Aliases
CG5483, CT17358, Dmel\CG5483, Dmel_CG5483, LRRK, LRRK2, Lrrk2, dLRRK, dLRRK2, dLrrk, lrrk
Chromosome Location
Chr.3R: 20635480 - 20644279 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038816
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104377.2 NP_001097847.1 5 - 6 3961 75
NM_001275842.1 NP_001262771.1 5 - 6 3961 75
NM_001275843.1 NP_001262772.1 6 - 7 4234 75
GenBank mRNA DQ494533.1 - 5 - 6 3872 75

View Full Product Details