Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syp
Entrez Gene ID
42460
Gene Name
Syncrip
Gene Aliases
AI945337, CG17838, Dmel\CG17838, Dmel_CG17838, anon-WO0118547.613, cg17838, l(3)03806, syp
Chromosome Location
Chr.3R: 20756729 - 20810726 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038826
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275848.1 NP_001262777.1 5 - 6 1156 109
NM_001275850.1 NP_001262779.1 6 - 7 1417 109
NM_169928.3 NP_732558.2 5 - 6 1856 109
GenBank mRNA AY058477.1 - 5 - 6 1705 109

View Full Product Details