Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RhoGAP93B
- Entrez Gene ID
-
42496
- Gene Name
- Rho GTPase activating protein at 93B
- Gene Aliases
-
CG3421, CrGAP, Dmel\CG3421, Dmel_CG3421, RhoGAP, Vilse, vilse
- Chromosome Location
-
Chr.3R: 21083043 - 21097827 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038853
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142682.2 |
NP_650939.2 |
4 - 5 |
1135 |
70 |
GenBank mRNA |
BT053685.1 |
- |
4 - 5 |
1113 |
70 |
View Full Product Details