Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snmp1
Entrez Gene ID
42514
Gene Name
Sensory neuron membrane protein 1
Gene Aliases
CG7000, Dmel\CG7000, Dmel_CG7000, SNMP, SNMP1, Snmp, snmp, snmp1
Chromosome Location
Chr.3R: 21165167 - 21168205 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260004
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275874.1 NP_001262803.1 1 - 2 188 101
NM_142696.3 NP_650953.1 1 - 2 188 101
GenBank mRNA BT024286.1 - 1 - 2 188 101

View Full Product Details