Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gld2
Entrez Gene ID
42602
Gene Name
GLD2 poly(A) polymerase
Gene Aliases
CG5732, Dm1, DmCG5732, DmGLD2, Dmel\CG5732, Dmel_CG5732, GLD-2, GLD2, TUTase2
Chromosome Location
Chr.3R: 22211824 - 22217415 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038934
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275900.1 NP_001262829.1 2 - 3 3049 68
NM_142755.4 NP_651012.2 1 - 2 2744 68

View Full Product Details