Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Takl2
Entrez Gene ID
42692
Gene Name
Tak1-like 2
Gene Aliases
CG4803, Dmel\CG4803, Dmel_CG4803, TAK, takl2
Chromosome Location
Chr.3R: 22732356 - 22733851 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039015
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_142833.2 NP_651090.2 1 - 2 82 66
GenBank mRNA BT132819.1 - 1 - 2 82 66

View Full Product Details