Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Miro
Entrez Gene ID
42845
Gene Name
Mitochondrial Rho
Gene Aliases
B682, CG5410, DMiro, DmMiro, Dmel\CG5410, Dmel_CG5410, Q8IMX7-1, Q8IMX7-2, dMiro, dmiro, miro
Chromosome Location
Chr.3R: 24034362 - 24038847 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039140
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275966.1 NP_001262895.1 4 - 5 3122 68
NM_142948.4 NP_651205.2 3 - 4 2067 68

View Full Product Details