Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mld
Entrez Gene ID
2768685
Gene Name
molting defective
Gene Aliases
CG13620, CG31312, CG33343, CG34100, CG9469, DTS-3, Dmel\CG34100, Dmel_CG34100, L(3)3[DTS], Mld, l(3)3[DTS], l(3)DTS3
Chromosome Location
Chr.3R: 24585653 - 24623899 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263490
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043289.2 NP_001036754.1 3 - 4 5171 61
NM_001043290.2 NP_001036755.2 3 - 4 5168 61
NM_001260360.1 NP_001247289.1 3 - 4 5171 61

View Full Product Details