Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- niki
- Entrez Gene ID
-
42959
- Gene Name
- nimA-like kinase
- Gene Aliases
-
CG10951, Dmel\CG10951, Dmel_CG10951, NEK1, Niki
- Chromosome Location
-
Chr.3R: 24822401 - 24825648 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0045980
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143036.2 |
NP_651293.1 |
7 - 8 |
2415 |
71 |
View Full Product Details