Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nmnat
Entrez Gene ID
42987
Gene Name
Nicotinamide mononucleotide adenylyltransferase
Gene Aliases
3R4, AAL90149, CG13645, DmNMNAT, DmNmnat, Dmel\CG13645, Dmel_CG13645, NMNAT, Nmnat1, NmnatB, dNmnat, nmnat
Chromosome Location
Chr.3R: 24945237 - 24947400 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039254
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276013.2 NP_001262942.1 1 - 2 182 63
NM_001369970.1 NP_001356924.1 1 - 2 182 63
NM_143058.1 NP_651315.2 1 - 2 218 63
NM_170185.1 NP_733064.2 1 - 2 218 63
GenBank mRNA AY089411.1 - 1 - 2 145 63
BT030750.1 - 1 - 2 217 63

View Full Product Details