Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RASSF8
- Entrez Gene ID
-
43096
- Gene Name
- Ras association domain family member 8
- Gene Aliases
-
Boa, CG5053, Dmel\CG5053, Dmel_CG5053, dRASSF8, dmRASSF7/8, rassf8
- Chromosome Location
-
Chr.3R: 25661795 - 25665640 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261986
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143154.4 |
NP_651411.1 |
3 - 4 |
1790 |
69 |
GenBank mRNA |
AY047500.1 |
- |
3 - 4 |
1769 |
69 |
View Full Product Details