Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLS
Entrez Gene ID
43104
Gene Name
Cardiolipin synthase
Gene Aliases
CG4774, Dmel\CG4774, Dmel_CG4774
Chromosome Location
Chr.3R: 25685236 - 25687370 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039360
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276040.1 NP_001262969.1 2 - 3 668 70
NM_143161.2 NP_651418.1 2 - 3 716 70
NM_170237.2 NP_733116.1 2 - 3 816 70
NM_170238.2 NP_733117.1 2 - 3 752 70
GenBank mRNA AY113254.1 - 2 - 3 816 70

View Full Product Details