Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TwdlC
- Entrez Gene ID
-
43245
- Gene Name
- TweedleC
- Gene Aliases
-
CG14254, Dmel\CG14254, Dmel_CG14254
- Chromosome Location
-
Chr.3R: 26994832 - 26996442 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039469
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143262.3 |
NP_651519.1 |
1 - 2 |
236 |
95 |
View Full Product Details