Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nep5
Entrez Gene ID
43255
Gene Name
Neprilysin 5
Gene Aliases
CG6265, DmeNEP5, Dmel\CG6265, Dmel_CG6265, NEP5, nep5
Chromosome Location
Chr.3R: 27057908 - 27064199 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039478
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104470.2 NP_001097940.1 1 - 2 496 60
NM_001300625.1 NP_001287554.1 1 - 2 917 60
NM_143270.3 NP_651527.3 1 - 2 496 60
NM_170307.2 NP_733186.2 1 - 2 496 60
GenBank mRNA BT001312.1 - 1 - 2 496 60

View Full Product Details