Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eater
Entrez Gene ID
43262
Gene Name
CG6124-PC
Gene Aliases
C901-like, CG6124, CT19227, Dmel\CG6124, Dmel_CG6124, Eater
Chromosome Location
Chr.3R: 27095867 - 27099679 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0243514
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_143276.3 NP_651533.3 3 - 4 3187 123

View Full Product Details