Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gr98a
- Entrez Gene ID
-
43305
- Gene Name
- Gustatory receptor 98a
- Gene Aliases
-
98B.4, CG13976, Dmel\CG13976, Dmel_CG13976, GR98B.4, GR98a, Gr98B4
- Chromosome Location
-
Chr.3R: 27487927 - 27489968 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039520
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143307.2 |
NP_651564.1 |
1 - 2 |
1097 |
87 |
GenBank mRNA |
BT050571.1 |
- |
1 - 2 |
921 |
87 |
View Full Product Details