Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ALiX
- Entrez Gene ID
-
43330
- Gene Name
- ALG-2 interacting protein X
- Gene Aliases
-
AIP1, ALIX, Aip1, Alix, CG12876, DAIP1, Dmel\CG12876, Dmel_CG12876, Snf7, alix
- Chromosome Location
-
Chr.3R: 27696466 - 27700110 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0086346
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143325.4 |
NP_651582.1 |
1 - 2 |
339 |
89 |
GenBank mRNA |
AY069534.1 |
- |
1 - 2 |
437 |
89 |
View Full Product Details