Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gp93
- Entrez Gene ID
-
43354
- Gene Name
- Glycoprotein 93
- Gene Aliases
-
CG5520, CT17486, Dmel\CG5520, Dmel_CG5520, GP93, HSP90, HSP90B1, Hsp90, Q9VAY2, dGRP94, dHSP90, gp93, p93
- Chromosome Location
-
Chr.3R: 27948711 - 27952151 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039562
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143344.3 |
NP_651601.1 |
4 - 5 |
1617 |
67 |
GenBank mRNA |
AY075384.1 |
- |
4 - 5 |
1583 |
67 |
View Full Product Details