Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Moca-cyp
Entrez Gene ID
43374
Gene Name
CG1866-PA
Gene Aliases
BEST:LD38313, CG1866, Dmel\CG1866, Dmel_CG1866, anon-WO0147981.13, anon-WO0147981.4, anon-WO0147981.5, moca
Chromosome Location
Chr.3R: 28340334 - 28345169 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039581
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170287.3 NP_001163758.1 6 - 7 1856 71
NM_143361.6 NP_651618.3 7 - 8 1689 71
NM_170367.3 NP_733246.1 7 - 8 1131 71
GenBank mRNA AY051595.1 - 7 - 8 1670 71

View Full Product Details