Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
beat-VI
Entrez Gene ID
43377
Gene Name
beaten path VI
Gene Aliases
Beat-VI, CG12089, CG14064, Dmel\CG14064, Dmel_CG14064, beat, beat VI
Chromosome Location
Chr.3R: 28418208 - 28474814 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039584
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276106.1 NP_001263035.1 4 - 5 993 92
NM_143362.3 NP_651619.1 4 - 5 993 92
GenBank mRNA AY052386.2 - 4 - 5 990 92
BT088757.1 - 4 - 5 993 92

View Full Product Details