Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ssl2
- Entrez Gene ID
-
43424
- Gene Name
- Strictosidine synthase-like 2
- Gene Aliases
-
CG11833, DmSSl2, Dmel\CG11833, Dmel_CG11833, SSl2
- Chromosome Location
-
Chr.3R: 29030951 - 29032642 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041780
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143399.3 |
NP_651656.2 |
1 - 2 |
231 |
63 |
View Full Product Details