Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
alph
Entrez Gene ID
43481
Gene Name
alphabet
Gene Aliases
Alph, CG1906, Dmel\CG1906, Dmel_CG1906, PP2Cb, SK3-1, pp2c99B
Chromosome Location
Chr.3R: 29591831 - 29617157 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086361
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_143444.4 NP_651701.1 3 - 4 1342 73
NM_170416.3 NP_733295.1 3 - 4 1342 73
NM_170417.3 NP_733296.1 3 - 4 1107 73
NM_170418.3 NP_733297.1 2 - 3 1183 73
NM_170419.3 NP_733298.1 3 - 4 1342 73
GenBank mRNA AY051685.1 - 3 - 4 1318 73

View Full Product Details