Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- alph
- Entrez Gene ID
-
43481
- Gene Name
- alphabet
- Gene Aliases
-
Alph, CG1906, Dmel\CG1906, Dmel_CG1906, PP2Cb, SK3-1, pp2c99B
- Chromosome Location
-
Chr.3R: 29591831 - 29617157 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0086361
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143444.4 |
NP_651701.1 |
5 - 6 |
1512 |
77 |
GenBank mRNA |
AY051685.1 |
- |
5 - 6 |
1488 |
77 |
View Full Product Details