Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ppk21
Entrez Gene ID
43485
Gene Name
pickpocket 21
Gene Aliases
CG12048, Dmel\CG12048, Dmel_CG12048, PPK21, Ppk21
Chromosome Location
Chr.3R: 29664779 - 29666783 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039675
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276144.1 NP_001263073.1 2 - 3 756 74
NM_143447.2 NP_651704.2 2 - 3 762 74

View Full Product Details