Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ppk20
Entrez Gene ID
43486
Gene Name
pickpocket 20
Gene Aliases
CG7577, Dmel\CG7577, Dmel_CG7577, PPK20
Chromosome Location
Chr.3R: 29666814 - 29668877 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039676
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276145.1 NP_001263074.1 6 - 7 1422 66
NM_001276146.1 NP_001263075.1 6 - 7 1425 66
NM_143448.2 NP_651705.2 5 - 6 1491 66

View Full Product Details