Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tace
Entrez Gene ID
43558
Gene Name
CG7908-PA
Gene Aliases
CG7908, CT23908, DTACE, Dmel\CG7908, Dmel_CG7908, TACE, TNF-alpha converting enzyme
Chromosome Location
Chr.3R: 30000530 - 30003678 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039734
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_143502.4 NP_651759.4 5 - 6 1858 70
NM_170455.3 NP_733334.1 7 - 8 1944 70
GenBank mRNA AY212802.1 - 7 - 8 1920 70
BT003184.1 - 7 - 8 1920 70

View Full Product Details