Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RpS28a
Entrez Gene ID
43564
Gene Name
Ribosomal protein S28a
Gene Aliases
CG15527, Dmel\CG15527, Dmel_CG15527
Chromosome Location
Chr.3R: 30022345 - 30022648 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039739
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_143507.2 NP_651764.1 1 - 1 98 78
GenBank mRNA BT100071.2 - 1 - 1 48 78
KX531288.1 - 1 - 1 48 78

View Full Product Details