Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aralar1
Entrez Gene ID
43616
Gene Name
Aralar1
Gene Aliases
Aralar, BcDNA:GH07418, CABP1, CG2139, DmAralar, Dmel\CG2139, Dmel_CG2139, aralar
Chromosome Location
Chr.3R: 30445635 - 30455867 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028646
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260439.1 NP_001247368.1 2 - 3 371 73
NM_001276178.1 NP_001263107.1 2 - 3 332 73
NM_143538.3 NP_651795.2 2 - 3 332 73
NM_170485.2 NP_733364.1 2 - 3 371 73
NM_170486.2 NP_733365.1 2 - 3 332 73
NM_170487.3 NP_733366.2 2 - 3 507 73
GenBank mRNA AY058654.1 - 2 - 3 297 73

View Full Product Details