Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ppk24
- Entrez Gene ID
-
43702
- Gene Name
- pickpocket 24
- Gene Aliases
-
CG15555, Dmel\CG15555, Dmel_CG15555, Ppk24
- Chromosome Location
-
Chr.3R: 31265730 - 31268511 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039839
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143603.2 |
NP_651860.2 |
2 - 3 |
460 |
59 |
View Full Product Details