Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- pont
- Entrez Gene ID
-
53439
- Gene Name
- pontin
- Gene Aliases
-
0229/05, CG4003, Dmel\CG4003, Dmel_CG4003, PON, PONT, Pont, Pontin, RUVBL1, Tip49, Tip49/Pontin, anon-WO0118547.698, dPon, dPontin, dRUVBL1, dpon, l(3)S022905, pon
- Chromosome Location
-
Chr.3R: 10262123 - 10263899 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0040078
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_144351.4 |
NP_652608.1 |
2 - 3 |
1238 |
55 |
GenBank mRNA |
AY061095.1 |
- |
2 - 3 |
1249 |
55 |
View Full Product Details