Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or82a
- Entrez Gene ID
-
318778
- Gene Name
- Odorant receptor 82a
- Gene Aliases
-
82a, CG31519, DOR61, DmOR82A, DmOr82a, Dmel\CG31519, Dmel_CG31519
- Chromosome Location
-
Chr.3R: 4256972 - 4258444 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041621
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_164323.1 |
NP_730794.1 |
5 - 6 |
1102 |
70 |
View Full Product Details