Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hph
Entrez Gene ID
40633
Gene Name
HIF prolyl hydroxylase
Gene Aliases
0303/04, 31543, CG1114, CG14665, CG2676, CG31543, CG44015, CR31541, Dm.egl-9, Dmel\CG44015, Dmel_CG44015, Fga, FgaB, PHD, anon-EST:Posey69, dHPH, dPHD, dPHD2, dmHPH, fatiga, fga, hph, l(3)02255, l(3)82Fe, l(3)S030304
Chromosome Location
Chr.3R: 5257037 - 5269507 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264785
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300242.1 NP_001287171.1 1 - 2 1090 63
NM_169036.2 NP_730906.1 1 - 2 1090 63
GenBank mRNA AY094896.1 - 1 - 2 1086 63

View Full Product Details