Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Itpr
- Entrez Gene ID
-
40664
- Gene Name
- Inositol 1,4,5,-trisphosphate receptor
- Gene Aliases
-
CG1063, DIP, DmInsP[[3R]], DmInsP[[3]]R, Dmel\CG1063, Dmel_CG1063, IP, IP3R, IP[[3]] receptor, IP[[3]]R, ITP-r83A, ITPR1, Ins3PR, InsP(3), InsP3, InsP3R, InsP[[3]], InsP[[3]]R, Insp3R, Ip3R, Itp-R83A, Itp-r, Itp-r-83A, Itp-r42B, Itp-r83A, Itp-r[1], anon-WO02059370.49, dIP3R, dIP[[3]]R, dip, itpr, itpr-83, l(3)05616, l(3)itpr, l(3)j5B4
- Chromosome Location
-
Chr.3R: 5517117 - 5539411 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0010051
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_169060.2 |
NP_730941.1 |
11 - 12 |
3978 |
72 |
View Full Product Details