Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pi4KIIalpha
Entrez Gene ID
40670
Gene Name
Phosphatidylinositol 4-kinase II alpha
Gene Aliases
CG2929, Dmel\CG2929, Dmel_CG2929, PI4KII, PI4KIIA, PI4KIIa, PI4KIIalpha, PI4KIIbeta, Pi4KII, Pi4KIIa
Chromosome Location
Chr.3R: 5557663 - 5563652 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037339
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_169063.4 NP_730944.2 2 - 3 526 67
NM_206432.3 NP_996154.1 2 - 3 499 67
GenBank mRNA AY102687.1 - 2 - 3 472 67

View Full Product Details