Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sec23
Entrez Gene ID
40694
Gene Name
Secretory 23
Gene Aliases
CG1250, COPII, Dmel\CG1250, Dmel_CG1250, Hau, SEC23, Sec23A, Sec23p, dSec23, dSec23p, dar2, dsec23, dsec23p, hau, sec23
Chromosome Location
Chr.3R: 5649065 - 5652629 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262125
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260003.1 NP_001246932.1 2 - 3 469 64
NM_001260004.1 NP_001246933.1 2 - 3 469 64
NM_001260005.2 NP_001246934.1 1 - 2 590 64
NM_001275365.1 NP_001262294.1 2 - 3 382 64
NM_001300255.1 NP_001287184.1 2 - 3 469 64
NM_169088.2 NP_730978.1 2 - 3 498 64
NM_169089.4 NP_730979.1 1 - 2 590 64

View Full Product Details