Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sec8
Entrez Gene ID
40712
Gene Name
Secretory 8
Gene Aliases
CG2095, Dmel\CG2095, Dmel_CG2095, Sec8p, dsec8, fun, sec8
Chromosome Location
Chr.3R: 5754932 - 5758997 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266672
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_169098.4 NP_730996.2 1 - 2 238 77
GenBank mRNA BT150369.1 - 1 - 2 200 77

View Full Product Details