Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pak
Entrez Gene ID
44039
Gene Name
p21-activated kinase
Gene Aliases
CG10295, D-Pak, DPAK, DPak, DPak1, DmPAK1, Dmel\CG10295, Dmel_CG10295, Dpak, Dpak1, PAK, PAK-1, PAK1, PAK2, PaK, Pak1, dPAK, dPAK1, dPak, dpak, dpak1, l(3)psg4, p65[PAK], pak, pak1, psg4
Chromosome Location
Chr.3R: 6350896 - 6361614 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0267698
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144541.2 NP_001138013.2 6 - 7 2790 80
NM_001275380.2 NP_001262309.1 6 - 7 2084 80
NM_001275381.2 NP_001262310.1 6 - 7 2497 80
NM_001275382.1 NP_001262311.1 6 - 7 2751 80
NM_001275383.1 NP_001262312.1 6 - 7 2492 80
NM_001275384.1 NP_001262313.1 6 - 7 2382 80
NM_079942.4 NP_524681.1 6 - 7 2089 80
NM_169136.3 NP_731073.1 6 - 7 2382 80
NM_169137.3 NP_731074.1 6 - 7 1985 80
GenBank mRNA BT010291.1 - 6 - 7 2350 80

View Full Product Details