Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SelR
Entrez Gene ID
41309
Gene Name
Selenoprotein R
Gene Aliases
BcDNA:RE36040, BcDNA:RH61230, CG6584, Dmel\CG6584, Dmel_CG6584, EP3340, MSRB, MsrB, dMsrB, selR
Chromosome Location
Chr.3R: 10863355 - 10868642 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0267376
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260107.2 NP_001247036.1 3 - 4 648 111
NM_169371.4 NP_731525.2 3 - 4 533 111
GenBank mRNA BT001621.1 - 3 - 4 454 111
BT120206.1 - 3 - 4 643 111

View Full Product Details