Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dpr17
Entrez Gene ID
41470
Gene Name
defective proboscis extension response 17
Gene Aliases
CG14737, CG14738, CG31361, CT34531, Dmel\CG31361, Dmel_CG31361, Dpr-17, Dpr17
Chromosome Location
Chr.3R: 12094876 - 12111420 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0051361
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144567.2 NP_001138039.1 5 - 6 2076 90
NM_001300368.1 NP_001287297.1 5 - 6 2892 90
NM_001300369.1 NP_001287298.1 5 - 6 2076 90
NM_169451.2 NP_731670.1 6 - 7 2898 90
NM_169452.3 NP_731671.1 5 - 6 2076 90
GenBank mRNA BT133437.1 - 6 - 7 2896 90

View Full Product Details