Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mthl12
- Entrez Gene ID
-
261599
- Gene Name
- methuselah-like 12
- Gene Aliases
-
CG32853, Dmel\CG32853, Dmel_CG32853, Mthl12
- Chromosome Location
-
Chr.3R: 13353673 - 13355413 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0045442
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_169508.2 |
NP_731791.2 |
1 - 2 |
515 |
126 |
View Full Product Details