Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ems
Entrez Gene ID
41697
Gene Name
empty spiracles
Gene Aliases
CG2988, Dmel\CG2988, Dmel_CG2988, E4, EMS, Ems, W13, ant
Chromosome Location
Chr.3R: 13901853 - 13904620 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000576
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_169560.2 NP_731868.1 1 - 2 1462 79
GenBank mRNA BT021310.1 - 1 - 2 1460 79

View Full Product Details