Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sap47
Entrez Gene ID
41938
Gene Name
Synapse-associated protein 47kD
Gene Aliases
CG8884, Dmel\CG8884, Dmel_CG8884, SAP 47, SAP-47, SAP47, dsap-47, sap47
Chromosome Location
Chr.3R: 15935100 - 15962124 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013334
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001370053.1 NP_001356917.1 6 - 7 2065 71
NM_001370054.1 NP_001356918.1 7 - 8 1699 71
NM_169684.3 NP_732088.1 7 - 8 2115 71
NM_169685.3 NP_732089.1 8 - 9 1723 71
NM_169686.3 NP_732090.1 8 - 9 2051 71
NM_169687.3 NP_732091.1 8 - 9 1869 71
NM_169689.3 NP_732093.1 7 - 8 1699 71
GenBank mRNA AY051871.1 - 7 - 8 2063 71

View Full Product Details