Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fas1
Entrez Gene ID
42025
Gene Name
Fasciclin 1
Gene Aliases
CG6588, DROFAS, DROFAS1, DROFASI, Dmel\CG6588, Dmel_CG6588, FGM222E05, Fas I, FasI, MRE24, fas I, fas-I, fas1, fasI, gasI
Chromosome Location
Chr.3R: 16625238 - 16639771 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0285925
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104344.2 NP_001097814.1 6 - 7 922 81
NM_001144593.2 NP_001138065.1 5 - 6 830 81
NM_001144594.2 NP_001138066.1 5 - 6 883 81
NM_001275716.1 NP_001262645.1 5 - 6 883 81
NM_169724.2 NP_732164.1 5 - 6 883 81
NM_169725.2 NP_732165.1 5 - 6 830 81
NM_169726.2 NP_732166.1 5 - 6 830 81
GenBank mRNA BT044209.1 - 5 - 6 861 81
M20545.1 - 5 - 6 809 81

View Full Product Details