Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dscam3
Entrez Gene ID
42103
Gene Name
Down syndrome cell adhesion molecule 3
Gene Aliases
CG12536, CG31190, CG8106, CT33960, Dmel\CG31190, Dmel_CG31190, Dscam-like
Chromosome Location
Chr.3R: 17473935 - 17505667 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261046
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104355.2 NP_001097825.1 17 - 18 3116 75
NM_001104356.2 NP_001097826.1 17 - 18 2639 75
NM_169761.2 NP_732242.2 17 - 18 2639 75
NM_206504.2 NP_996226.2 17 - 18 2639 75
GenBank mRNA BT014656.1 - 17 - 18 2587 75

View Full Product Details