Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpo
Entrez Gene ID
45840
Gene Name
couch potato
Gene Aliases
1824, 3603, 5339, 6015, BcDNA:AT31405, CG12349, CG18434, CG18435, CG31243, CG42457, CG43738, CPO, Cpo, Dmel\CG43738, Dmel_CG43738, E(sda)N, Line N, ORE-17, cg18435, l(3)01432, l(3)6015, l(3)j4A1, l(3)j4D4, l(3)j7E2, l(3)j9B3, l(3)rJ553, l(3)s2336
Chromosome Location
Chr.3R: 17919832 - 18018892 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263995
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014631.3 NP_001014631.3 5 - 6 3180 97
NM_001014632.4 NP_001014632.3 5 - 6 2069 97
NM_001170169.3 NP_001163640.2 5 - 6 1836 97
NM_001275759.1 NP_001262688.1 5 - 6 1996 97
NM_001275760.1 NP_001262689.1 5 - 6 2069 97
NM_001275761.1 NP_001262690.1 5 - 6 2124 97
NM_001275762.1 NP_001262691.1 5 - 6 2124 97
NM_001275763.1 NP_001262692.1 5 - 6 2069 97
NM_080105.4 NP_524844.6 5 - 6 2069 97
NM_169781.4 NP_732282.6 5 - 6 3180 97
NM_169782.3 NP_732283.5 5 - 6 2124 97
NM_169783.4 NP_732284.5 5 - 6 2069 97

View Full Product Details